4-Methoxy-3-(trifluoromethoxy)benzylamine

Lieferant: Apollo Scientific

PC302604-1G PC302604-5G
APOSPC302604-1GEA 0 EUR
APOSPC302604-1G APOSPC302604-5G
4-Methoxy-3-(trifluoromethoxy)benzylamine
4-Methoxy-3-(trifluoromethoxy)benzylamine
Formel: C₉H₁₀F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18398479
CAS-Nummer: 1261865-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR