4-Methoxy-3-(trifluormethyl)benzolsulfonylchlorid 98%
Lieferant: Apollo Scientific
PC302605-1G
PC302605-5G
PC302605-10G
APOSPC302605-1GEA
171
EUR
APOSPC302605-1G
APOSPC302605-5G
APOSPC302605-10G
4-Methoxy-3-(trifluormethyl)benzolsulfonylchlorid 98%
4-Methoxy-3-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₈H₆ClF₃O₃S Schmelzpunkt: 52…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07774167 CAS-Nummer: 694514-21-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...