4-Methoxy-3-(trifluoromethoxy)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC302607-1G
APOSPC302607-1GEA 0 EUR
APOSPC302607-1G
4-Methoxy-3-(trifluoromethoxy)benzenesulphonyl chloride
4-Methoxy-3-(trifluoromethoxy)benzenesulphonyl chloride
Formel: C₈H₆ClF₃O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD18398444
CAS-Nummer: 956468-19-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR