5-(Difluormethoxy)-3-fluorbenzylbromid 95%

Lieferant: Apollo Scientific
Gefahr

PC302611-1G PC302611-5G
APOSPC302611-1GEA 116 EUR
APOSPC302611-1G APOSPC302611-5G
5-(Difluormethoxy)-3-fluorbenzylbromid 95%
5-(Difluormethoxy)-3-fluorbenzylbromid
Formel: C₈H₆BrF₃O
Molecular Weight: 255.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832362
CAS-Nummer: 1017779-39-3
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR