5-Chlor-2-(trifluormethoxy)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC302614-1G PC302614-5G
APOSPC302614-1GEA 178 EUR
APOSPC302614-1G APOSPC302614-5G
5-Chlor-2-(trifluormethoxy)benzylbromid 98%
5-Chlor-2-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD11519358
CAS-Nummer: 1092461-19-2
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR