5-Chloro-2-(trifluoromethyl)-DL-phenylalanine

Lieferant: Apollo Scientific

PC302615-1G PC302615-5G PC302615-10G
APOSPC302615-1GEA 0 EUR
APOSPC302615-1G APOSPC302615-5G APOSPC302615-10G
5-Chloro-2-(trifluoromethyl)-DL-phenylalanine
5-Chloro-2-(trifluoromethyl)-DL-phenylalanine
Formel: C₁₀H₉ClF₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832385
CAS-Nummer: 1256482-62-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR