5-Fluor-2-methoxybenzylamin 97%

Lieferant: Apollo Scientific

PC302621-1G PC302621-5G PC302621-10G PC302621-25G
APOSPC302621-1GEA 0 EUR
APOSPC302621-1G APOSPC302621-5G APOSPC302621-10G APOSPC302621-25G
5-Fluor-2-methoxybenzylamin 97%
5-Fluor-2-methoxybenzylamin
Formel: C₈H₁₀FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD06213625
CAS-Nummer: 148870-38-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR