5-Fluoro-2-methoxy-DL-phenylalanine
Lieferant: Apollo Scientific
PC302622-1G
APOSPC302622-1GEA
0
EUR
APOSPC302622-1G
5-Fluoro-2-methoxy-DL-phenylalanine
5-Fluoro-2-methoxy-DL-phenylalanine
Formel:
C₁₀H₁₂FNO₃ Schmelzpunkt: 251 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05227952 CAS-Nummer: 603105-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...