5-Fluoro-2-methyl-DL-phenylglycine

Lieferant: Apollo Scientific

PC302625-1G
APOSPC302625-1GEA 159 EUR
APOSPC302625-1G
5-Fluoro-2-methyl-DL-phenylglycine
5-Fluoro-2-methyl-DL-phenylglycine
Formel: C₉H₁₀FNO₂
Molecular Weight: 183,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116047
CAS-Nummer: 1039365-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR