5-Methyl-2-(trifluoromethyl)-DL-phenylalanine

Lieferant: Apollo Scientific

PC302626-1G PC302626-5G PC302626-10G
APOSPC302626-1GEA 224 EUR
APOSPC302626-1G APOSPC302626-5G APOSPC302626-10G
5-Methyl-2-(trifluoromethyl)-DL-phenylalanine
5-Methyl-2-(trifluoromethyl)-DL-phenylalanine
Formel: C₁₁H₁₂F₃NO₂
Molecular Weight: 247.214 g/mol
Schmelzpunkt: 236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832357
CAS-Nummer: 1256482-64-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR