6-Chloro-2,3-difluoro-DL-phenylalanine

Lieferant: Apollo Scientific

Synonyme: 2-Amino-3-(6-chloro-2,3-difluorophenyl)propanoic acid

PC302630-1G
APOSPC302630-1GEA 116 EUR
APOSPC302630-1G
6-Chloro-2,3-difluoro-DL-phenylalanine
6-Chloro-2,3-difluoro-DL-phenylalanine
Formel: C₉H₈ClF₂NO₂
Molecular Weight: 235,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201054
CAS-Nummer: 1706428-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR