2,3-Difluoro-6-methoxybenzyl alcohol 97%

Lieferant: Apollo Scientific

PC302660-10G PC302660-5G PC302660-1G
APOSPC302660-10GEA 337 EUR
APOSPC302660-10G APOSPC302660-5G APOSPC302660-1G
2,3-Difluoro-6-methoxybenzyl alcohol 97%
2,3-Difluoro-6-methoxybenzyl alcohol
Formel: C₈H₈F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04116021
CAS-Nummer: 773871-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR