2,6-Difluoro-3-methoxybenzoyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302667-250MG
APOSPC302667-250MGEA
0
EUR
APOSPC302667-250MG
2,6-Difluoro-3-methoxybenzoyl chloride 97%
2,6-Difluoro-3-methoxybenzoyl chloride
Formel:
C₈H₅ClF₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115903 CAS-Nummer: 886498-40-4 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...