2-Fluor-5-methyl-4-(trifluormethyl)benzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC302687-1G PC302687-5G
APOSPC302687-1GEA 313 EUR
APOSPC302687-1G APOSPC302687-5G
2-Fluor-5-methyl-4-(trifluormethyl)benzonitril 98%
2-Fluor-5-methyl-4-(trifluormethyl)benzonitril
Formel: C₉H₅F₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD19687195
CAS-Nummer: 1323966-20-6
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR