2-Fluor-5-methyl-4-(trifluormethyl)benzylamin 97%

Lieferant: Apollo Scientific

PC302688-1G
APOSPC302688-1GEA 190 EUR
APOSPC302688-1G
2-Fluor-5-methyl-4-(trifluormethyl)benzylamin 97%
2-Fluor-5-methyl-4-(trifluormethyl)benzylamin
Formel: C₉H₉F₄N
Molecular Weight: 207,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687196
CAS-Nummer: 1323966-36-4
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR