2,5-Bis(trifluormethyl)-6-methoxybenzaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC302691-10G
PC302691-5G
PC302691-1G
APOSPC302691-10GEA
237
EUR
APOSPC302691-10G
APOSPC302691-5G
APOSPC302691-1G
2,5-Bis(trifluormethyl)-6-methoxybenzaldehyd 97%
2,5-Bis(trifluormethyl)-6-methoxybenzaldehyd
Formel:
C₁₀H₆F₆O₂ Schmelzpunkt: 36…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660316 CAS-Nummer: 180134-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...