2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid 98%

Lieferant: Apollo Scientific

PC302692-10G PC302692-5G PC302692-1G PC302692-25G PC302692-100G
APOSPC302692-10GEA 0 EUR
APOSPC302692-10G APOSPC302692-5G APOSPC302692-1G APOSPC302692-25G APOSPC302692-100G
2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid 98%
2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid
Formel: C₁₀H₆F₆O₃
Schmelzpunkt: 156…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00728738
CAS-Nummer: 180134-15-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR