2-Methoxy-5-(trifluormethoxy)benzylamin 95%

Lieferant: Apollo Scientific

PC302693-5G PC302693-1G
APOSPC302693-5GEA 132 EUR
APOSPC302693-5G APOSPC302693-1G
2-Methoxy-5-(trifluormethoxy)benzylamin 95%
2-Methoxy-5-(trifluormethoxy)benzylamin
Formel: C₉H₁₀F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04972912
CAS-Nummer: 771582-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR