(2-Methyl-5-(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

PC302699-1G PC302699-5G PC302699-10G
APOSPC302699-1GEA 183 EUR
APOSPC302699-1G APOSPC302699-5G APOSPC302699-10G
(2-Methyl-5-(trifluormethyl)phenyl)essigsäure 98%
(2-Methyl-5-(trifluormethyl)phenyl)essigsäure
Formel: C₁₀H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832286
CAS-Nummer: 1000517-01-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR