3,5-Difluor-2-methylbenzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC302701-1G
APOSPC302701-1GEA 136 EUR
APOSPC302701-1G
3,5-Difluor-2-methylbenzonitril 98%
3,5-Difluor-2-methylbenzonitril
Formel: C₈H₅F₂N
Molecular Weight: 153,13 g/mol
Schmelzpunkt: 55…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09996934
CAS-Nummer: 1003708-74-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR