3-[4-Methyl-3-(trifluoromethyl)phenyl]propionic acid 98%

Lieferant: Apollo Scientific

PC302707-1G PC302707-5G PC302707-10G
APOSPC302707-1GEA 203 EUR
APOSPC302707-1G APOSPC302707-5G APOSPC302707-10G
3-[4-Methyl-3-(trifluoromethyl)phenyl]propionic acid 98%
3-[4-Methyl-3-(trifluoromethyl)phenyl]propionic acid
Formel: C₁₁H₁₁F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832295
CAS-Nummer: 1017778-20-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR