3-[5-Methyl-2-(trifluoromethyl)phenyl]propionic acid 98%

Lieferant: Apollo Scientific

PC302708-1G
APOSPC302708-1GEA 136 EUR
APOSPC302708-1G
3-[5-Methyl-2-(trifluoromethyl)phenyl]propionic acid 98%
3-[5-Methyl-2-(trifluoromethyl)phenyl]propionic acid
Formel: C₁₁H₁₁F₃O₂
Molecular Weight: 232,199 g/mol
Schmelzpunkt: 63…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832355
CAS-Nummer: 1017779-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR