3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetonitrile 97%

Lieferant: Apollo Scientific

PC302711-1G
APOSPC302711-1GEA 203 EUR
APOSPC302711-1G
3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetonitrile 97%
3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetonitrile
Formel: C₉H₄ClF₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD23795330
CAS-Nummer: 1431329-60-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR