3-Chloro-4-ethoxy-5-fluorobenzoyl chloride

Lieferant: Apollo Scientific

PC302714-1G
APOSPC302714-1GEA 60.9 EUR
APOSPC302714-1G
3-Chloro-4-ethoxy-5-fluorobenzoyl chloride
3-Chloro-4-ethoxy-5-fluorobenzoyl chloride
Formel: C₉H₇Cl₂FO₂
Molecular Weight: 237,055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832337
CAS-Nummer: 1017778-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR