3-Chloro-5-fluoro-4-methoxybenzoyl chloride

Lieferant: Apollo Scientific
Gefahr

PC302717-10G PC302717-5G
APOSPC302717-10GEA 0 EUR
APOSPC302717-10G APOSPC302717-5G
3-Chloro-5-fluoro-4-methoxybenzoyl chloride
3-Chloro-5-fluoro-4-methoxybenzoyl chloride
Formel: C₈H₅Cl₂FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04115865
CAS-Nummer: 886497-26-3
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR