3'-Ethoxy-2',4'-difluoracetophenon 98%
Lieferant: Apollo Scientific
Synonyme:
2,4-Difluoro-3-ethoxy acetophenone
PC302720-10G
PC302720-5G
PC302720-1G
APOSPC302720-10GEA
282
EUR
APOSPC302720-10G
APOSPC302720-5G
APOSPC302720-1G
3'-Ethoxy-2',4'-difluoracetophenon 98%
3'-Ethoxy-2',4'-difluoracetophenon
Formel:
C₁₀H₁₀F₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258734 CAS-Nummer: 1017778-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...