3-Ethoxy-2,6-difluoro-DL-phenylalanine

Lieferant: Apollo Scientific
Achtung

PC302723-1G
APOSPC302723-1GEA 136 EUR
APOSPC302723-1G
3-Ethoxy-2,6-difluoro-DL-phenylalanine
3-Ethoxy-2,6-difluoro-DL-phenylalanine
Formel: C₁₁H₁₃F₂NO₃
Molecular Weight: 245,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11519376
CAS-Nummer: 1260007-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR