3-Fluor-2-methoxybenzylalkohol 98%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-2-methoxybenzenemethanol

PC302726-5G
APOSPC302726-5GEA 148 EUR
APOSPC302726-5G
3-Fluor-2-methoxybenzylalkohol 98%
3-Fluor-2-methoxybenzylalkohol
Formel: C₈H₉FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09025369
CAS-Nummer: 303043-91-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR