3-Fluoro-5-methylbenzoyl chloride

Lieferant: Apollo Scientific

PC302727-10G PC302727-5G PC302727-1G
APOSPC302727-10GEA 250 EUR
APOSPC302727-10G APOSPC302727-5G APOSPC302727-1G
3-Fluoro-5-methylbenzoyl chloride
3-Fluoro-5-methylbenzoyl chloride
Formel: C₈H₆ClFO
Molecular Weight: 172.584 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115884
CAS-Nummer: 886497-77-4
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR