3-Methoxy-2-(trifluormethyl)benzylamin 98%

Lieferant: Apollo Scientific
Gefahr

PC302730-1G
APOSPC302730-1GEA 202 EUR
APOSPC302730-1G
3-Methoxy-2-(trifluormethyl)benzylamin 98%
3-Methoxy-2-(trifluormethyl)benzylamin
Formel: C₉H₁₀F₃NO
Molecular Weight: 205,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398592
CAS-Nummer: 1261647-71-5
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR