3'-Methyl-5'-(trifluormethoxy)acetophenon 98%

Lieferant: Apollo Scientific

PC302732-1G
APOSPC302732-1GEA 180 EUR
APOSPC302732-1G
3'-Methyl-5'-(trifluormethoxy)acetophenon 98%
3'-Methyl-5'-(trifluormethoxy)acetophenon
Formel: C₁₆H₁₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09025377
CAS-Nummer: 916420-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR