4-Chloro-3-fluoro-5-(trifluoromethyl)phenylacetonitrile 97%

Lieferant: Apollo Scientific

PC302743-1G
APOSPC302743-1GEA 224 EUR
APOSPC302743-1G
4-Chloro-3-fluoro-5-(trifluoromethyl)phenylacetonitrile 97%
4-Chloro-3-fluoro-5-(trifluoromethyl)phenylacetonitrile
Formel: C₉H₄ClF₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD23795335
CAS-Nummer: 1431329-62-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR