6-Chloro-2,3-difluorobenzoyl chloride 98%
Lieferant: Apollo Scientific
Synonyme:
Benzoyl chloride, 6-chloro-2,3-difluoro-
PC302763-1G
APOSPC302763-1GEA
69.6
EUR
APOSPC302763-1G
6-Chloro-2,3-difluorobenzoyl chloride 98%
6-Chloro-2,3-difluorobenzoyl chloride
Formel:
C₇H₂Cl₂F₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD12922578 CAS-Nummer: 1208078-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...