2-Fluoro-4-methylbenzamide 97%

Lieferant: Apollo Scientific

PC302765-1G
APOSPC302765-1GEA 48 EUR
APOSPC302765-1G
2-Fluoro-4-methylbenzamide 97%
2-Fluoro-4-methylbenzamide
Formel: C₈H₈FNO
Molecular Weight: 153.154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660328
CAS-Nummer: 886761-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR