3-Ethoxy-2,4-difluorbenzylamin 95%

Lieferant: Apollo Scientific

PC302775-1G
APOSPC302775-1GEA 277 EUR
APOSPC302775-1G
3-Ethoxy-2,4-difluorbenzylamin 95%
3-Ethoxy-2,4-difluorbenzylamin
Formel: C₉H₁₁F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09258733
CAS-Nummer: 1017778-36-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR