4-Chlor-3-fluorbenzylamin 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (4-Chloro-3-fluorophenyl)methanamine

PC302780-5G PC302780-1G
APOSPC302780-5GEA 48 EUR
APOSPC302780-5G APOSPC302780-1G
4-Chlor-3-fluorbenzylamin 97%
4-Chlor-3-fluorbenzylamin
Formel: C₇H₇ClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD04116008
CAS-Nummer: 72235-58-6
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR