4-Ethoxy-3-(trifluoromethyl)phenylacetic acid 98%

Lieferant: Apollo Scientific

PC302784-1G
APOSPC302784-1GEA 203 EUR
APOSPC302784-1G
4-Ethoxy-3-(trifluoromethyl)phenylacetic acid 98%
4-Ethoxy-3-(trifluoromethyl)phenylacetic acid
Formel: C₁₁H₁₁F₃O₃
Molecular Weight: 248,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13196586
CAS-Nummer: 1206593-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR