4-Methoxy-2-(trifluoromethoxy)benzamide 97%
Lieferant: Apollo Scientific
PC302786-1G
APOSPC302786-1GEA
144
EUR
APOSPC302786-1G
4-Methoxy-2-(trifluoromethoxy)benzamide 97%
4-Methoxy-2-(trifluoromethoxy)benzamide
Formel:
C₉H₈F₃NO₃ Molecular Weight: 235,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660287 CAS-Nummer: 886502-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...