2,3-Difluor-4-methoxybenzylbromid

Lieferant: Apollo Scientific

PC302787-10G PC302787-5G PC302787-1G
APOSPC302787-10GEA 372 EUR
APOSPC302787-10G APOSPC302787-5G APOSPC302787-1G
2,3-Difluor-4-methoxybenzylbromid
2,3-Difluor-4-methoxybenzylbromid
Formel: C₈H₇BrF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD04115985
CAS-Nummer: 689254-23-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR