2,3-Difluor-4-methoxybenzylbromid
Lieferant: Apollo Scientific
PC302787-10G
PC302787-5G
PC302787-1G
APOSPC302787-10GEA
372
EUR
APOSPC302787-10G
APOSPC302787-5G
APOSPC302787-1G
2,3-Difluor-4-methoxybenzylbromid
2,3-Difluor-4-methoxybenzylbromid
Formel:
C₈H₇BrF₂O Molecular Weight: 237.04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115985 CAS-Nummer: 689254-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...