2-Chloro-5-(pentafluorothio)benzyl alcohol 97%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-5-(pentafluorothio)benzyl alcohol
PC302795-1G
APOSPC302795-1GEA
308
EUR
APOSPC302795-1G
2-Chloro-5-(pentafluorothio)benzyl alcohol 97%
2-Chloro-5-(pentafluorothio)benzyl alcohol
Formel:
C₇H₆ClF₅OS Molecular Weight: 268,83 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22201048 CAS-Nummer: 1373920-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...