2-Fluoro-4-(pentafluorothio)phenylacetonitrile 98%

Lieferant: Apollo Scientific

PC302800-100MG
APOSPC302800-100MGEA 0 EUR
APOSPC302800-100MG
2-Fluoro-4-(pentafluorothio)phenylacetonitrile 98%
2-Fluoro-4-(pentafluorothio)phenylacetonitrile
Formel: C₈H₅F₆NS
Lagertemperatur: Raumtemperatur
MDL: MFCD16652508
CAS-Nummer: 1240256-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR