2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

PC302805-1G
APOSPC302805-1GEA 87 EUR
APOSPC302805-1G
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoyl chloride 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoyl chloride
Formel: C₉H₅ClF₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22201039
CAS-Nummer: 1373920-78-5
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR