2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetonitrile 98%

Lieferant: Apollo Scientific

PC302808-1G PC302808-5G
APOSPC302808-1GEA 0 EUR
APOSPC302808-1G APOSPC302808-5G
2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetonitrile 98%
2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetonitrile
Formel: C₁₀H₇F₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD19687199
CAS-Nummer: 1323966-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR