2-Methoxy-4-(trifluormethoxy)benzylamin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302809-1G
APOSPC302809-1GEA
112
EUR
APOSPC302809-1G
2-Methoxy-4-(trifluormethoxy)benzylamin 98%
2-Methoxy-4-(trifluormethoxy)benzylamin
Formel:
C₉H₁₀F₃NO₂ Molecular Weight: 221.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660330 CAS-Nummer: 886761-62-2 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...