2-Methoxy-4-(trifluormethyl)benzylbromid
Lieferant: Apollo Scientific
|
Gefahr
|
PC302810-1G
PC302810-5G
PC302810-25G
APOSPC302810-1GEA
58
EUR
APOSPC302810-1G
APOSPC302810-5G
APOSPC302810-25G
2-Methoxy-4-(trifluormethyl)benzylbromid
2-Methoxy-4-(trifluormethyl)benzylbromid
Formel:
C₉H₈BrF₃O Schmelzpunkt: 42…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115980 CAS-Nummer: 886500-59-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...