2'-Methoxy-5'-(trifluormethoxy)acetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302814-10G
PC302814-5G
PC302814-1G
APOSPC302814-10GEA
0
EUR
APOSPC302814-10G
APOSPC302814-5G
APOSPC302814-1G
2'-Methoxy-5'-(trifluormethoxy)acetophenon 98%
2'-Methoxy-5'-(trifluormethoxy)acetophenon
Formel:
C₁₀H₉F₃O₃ Schmelzpunkt: 45…47 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08062375 CAS-Nummer: 468074-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...