2-Methyl-5-(pentafluorthio)benzoesäure 98%

Lieferant: Apollo Scientific

PC302817-1G
APOSPC302817-1GEA 422 EUR
APOSPC302817-1G
2-Methyl-5-(pentafluorthio)benzoesäure 98%
2-Methyl-5-(pentafluorothio)benzoic acid
Formel: C₈H₇F₅O₂S
Molecular Weight: 262.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652477
CAS-Nummer: 623943-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR