2-Methyl-5-(trifluormethoxy)benzamid 97%

Lieferant: Apollo Scientific

PC302818-1G
APOSPC302818-1GEA 0 EUR
APOSPC302818-1G
2-Methyl-5-(trifluormethoxy)benzamid 97%
2-Methyl-5-(trifluormethoxy)benzamid
Formel: C₉H₈F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18399714
CAS-Nummer: 1261828-46-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR