3,5-Difluoro-2-methoxybenzoyl chloride
Lieferant: Apollo Scientific
PC302823-10G
PC302823-5G
APOSPC302823-10GEA
282
EUR
APOSPC302823-10G
APOSPC302823-5G
3,5-Difluoro-2-methoxybenzoyl chloride
3,5-Difluoro-2-methoxybenzoyl chloride
Formel:
C₈H₅ClF₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660199 CAS-Nummer: 886498-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...